1-[(2,4-Difluorophenoxy)methyl]-3,5-dimethyl-4-nitro-1H-pyrazole

Lieferant: Apollo Scientific

PC410287-1G
APOSPC410287-1GEA 386 EUR
APOSPC410287-1G
1-[(2,4-Difluorophenoxy)methyl]-3,5-dimethyl-4-nitro-1H-pyrazole
1-[(2,4-Difluorophenoxy)methyl]-3,5-dimethyl-4-nitro-1H-pyrazole
Formel: C₁₂H₁₁F₂N₃O₃
Molecular Weight: 283,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05861659
CAS-Nummer: 1001519-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR