[3-Cyclopropyl-5-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid
Lieferant: Apollo Scientific
PC410288-1G
APOSPC410288-1GEA
1080
EUR
APOSPC410288-1G
[3-Cyclopropyl-5-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid
[3-Cyclopropyl-5-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid
Formel:
C₉H₁₀F₂N₂O₂ Molecular Weight: 216.185 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05861720 CAS-Nummer: 1001519-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...