N-(2-Aminoethyl)-1-ethyl-3-(trifluoromethyl)-1H-pyrazole-5-carboxamide

Lieferant: Apollo Scientific

PC410289-1G
APOSPC410289-1GEA 647 EUR
APOSPC410289-1G
N-(2-Aminoethyl)-1-ethyl-3-(trifluoromethyl)-1H-pyrazole-5-carboxamide
N-(2-Aminoethyl)-1-ethyl-3-(trifluoromethyl)-1H-pyrazole-5-carboxamide
Formel: C₉H₁₃F₃N₄O
Molecular Weight: 250,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05862156
CAS-Nummer: 1001519-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR