4-Chloro-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

PC410290-1G
APOSPC410290-1GEA 647 EUR
APOSPC410290-1G
4-Chloro-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
4-Chloro-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
Formel: C₆H₄ClF₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03419816
CAS-Nummer: 1001519-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR