1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide
Lieferant: Apollo Scientific
PC410292-1G
APOSPC410292-1GEA
190
EUR
APOSPC410292-1G
1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide
1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide
Formel:
C₆H₇F₃N₄O Molecular Weight: 208,141 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419818 CAS-Nummer: 1001519-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...