4-Bromo-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide

Lieferant: Apollo Scientific

PC410293-1G
APOSPC410293-1GEA 386 EUR
APOSPC410293-1G
4-Bromo-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide
4-Bromo-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide
Formel: C₆H₆BrF₃N₄O
Molecular Weight: 287.037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419820
CAS-Nummer: 1001519-41-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR