{[4-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]sulphanyl}acetic acid

Lieferant: Apollo Scientific

PC410294-1G
APOSPC410294-1GEA 647 EUR
APOSPC410294-1G
{[4-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]sulphanyl}acetic acid
{[4-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]sulphanyl}acetic acid
Formel: C₁₃H₁₃F₃N₄O₂S
Molecular Weight: 346,328 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422187
CAS-Nummer: 1001560-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR