[5-Methyl-4-nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410295-1G
APOSPC410295-1GEA 993 EUR
APOSPC410295-1G
[5-Methyl-4-nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid
[5-Methyl-4-nitro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid
Formel: C₇H₆F₃N₃O₄
Molecular Weight: 253,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00297973
CAS-Nummer: 1001754-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR