2-Fluor-6-phenoxybenzonitril 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC4103-25G
PC4103-5G
PC4103-100G
APOSPC4103-25GEA
63.8
EUR
APOSPC4103-25G
APOSPC4103-5G
APOSPC4103-100G
2-Fluor-6-phenoxybenzonitril 95%
2-Fluor-6-phenoxybenzonitril
Formel:
C₁₃H₈FNO Schmelzpunkt: 90…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068201 CAS-Nummer: 175204-06-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...