4-Bromo-1-(2-fluorobenzyl)-1H-pyrazol-3-amine

Lieferant: Apollo Scientific

PC410300-1G
APOSPC410300-1GEA 516 EUR
APOSPC410300-1G
4-Bromo-1-(2-fluorobenzyl)-1H-pyrazol-3-amine
4-Bromo-1-(2-fluorobenzyl)-1H-pyrazol-3-amine
Formel: C₁₀H₉BrFN₃
Molecular Weight: 270,101 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02254032
CAS-Nummer: 1001757-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR