4-Bromo-1-(4-fluorobenzyl)-1H-pyrazol-3-amine
Lieferant: Apollo Scientific
PC410301-1G
APOSPC410301-1GEA
516
EUR
APOSPC410301-1G
4-Bromo-1-(4-fluorobenzyl)-1H-pyrazol-3-amine
4-Bromo-1-(4-fluorobenzyl)-1H-pyrazol-3-amine
Formel:
C₁₀H₉BrFN₃ Molecular Weight: 270,101 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02254037 CAS-Nummer: 1001757-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...