1-(Difluoromethyl)-1H-pyrazole-5-carbohydrazide
Lieferant: Apollo Scientific
PC410302-1G
APOSPC410302-1GEA
183
EUR
APOSPC410302-1G
1-(Difluoromethyl)-1H-pyrazole-5-carbohydrazide
1-(Difluoromethyl)-1H-pyrazole-5-carbohydrazide
Formel:
C₅H₆F₂N₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD04967153 CAS-Nummer: 1002032-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...