[5-Cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410303-1G
APOSPC410303-1GEA 828 EUR
APOSPC410303-1G
[5-Cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid
[5-Cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid
Formel: C₉H₁₀F₂N₂O₂
Molecular Weight: 216,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04967182
CAS-Nummer: 1002032-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR