[4-Chloro-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410304-1G
APOSPC410304-1GEA 647 EUR
APOSPC410304-1G
[4-Chloro-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid
[4-Chloro-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid
Formel: C₉H₉ClF₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04967193
CAS-Nummer: 1002032-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR