1-(2,3,4,6-Tetrafluorobenzyl)-1H-pyrazol-4-amine

Lieferant: Apollo Scientific

PC410305-1G
APOSPC410305-1GEA 516 EUR
APOSPC410305-1G
1-(2,3,4,6-Tetrafluorobenzyl)-1H-pyrazol-4-amine
1-(2,3,4,6-Tetrafluorobenzyl)-1H-pyrazol-4-amine
Formel: C₁₀H₇F₄N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04967237
CAS-Nummer: 1002032-97-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR