2-{[1-(4-Methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-5-yl]oxy}acetohydrazide
Lieferant: Apollo Scientific
PC410306-1G
APOSPC410306-1GEA
386
EUR
APOSPC410306-1G
2-{[1-(4-Methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-5-yl]oxy}acetohydrazide
2-{[1-(4-Methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-5-yl]oxy}acetohydrazide
Formel:
C₁₃H₁₃F₃N₄O₂ Molecular Weight: 314,263 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04967374 CAS-Nummer: 1002034-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...