1-(Difluoromethyl)-3-nitro-1H-pyrazole

Lieferant: Apollo Scientific

PC410307-1G
APOSPC410307-1GEA 168 EUR
APOSPC410307-1G
1-(Difluoromethyl)-3-nitro-1H-pyrazole
1-(Difluoromethyl)-3-nitro-1H-pyrazole
Formel: C₄H₃F₂N₃O₂
Molecular Weight: 163.082 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04967410
CAS-Nummer: 1002034-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR