1-(2-Chloro-6-fluorobenzyl)-4-nitro-1H-pyrazole
Lieferant: Apollo Scientific
PC410308-1G
APOSPC410308-1GEA
386
EUR
APOSPC410308-1G
1-(2-Chloro-6-fluorobenzyl)-4-nitro-1H-pyrazole
1-(2-Chloro-6-fluorobenzyl)-4-nitro-1H-pyrazole
Formel:
C₁₀H₇ClFN₃O₂ Molecular Weight: 255,633 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02575856 CAS-Nummer: 333444-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...