1-(Difluoromethyl)-5-methyl-1H-pyrazole

Lieferant: Apollo Scientific

PC410309-1G
APOSPC410309-1GEA 168 EUR
APOSPC410309-1G
1-(Difluoromethyl)-5-methyl-1H-pyrazole
1-(Difluoromethyl)-5-methyl-1H-pyrazole
Formel: C₅H₆F₂N₂
Molecular Weight: 132.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04968779
CAS-Nummer: 1004096-15-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR