1-[(3-Fluorophenoxy)methyl]-3,5-dimethyl-1H-pyrazol-4-amine

Lieferant: Apollo Scientific

PC410310-1G
APOSPC410310-1GEA 386 EUR
APOSPC410310-1G
1-[(3-Fluorophenoxy)methyl]-3,5-dimethyl-1H-pyrazol-4-amine
1-[(3-Fluorophenoxy)methyl]-3,5-dimethyl-1H-pyrazol-4-amine
Formel: C₁₂H₁₄FN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD04967714
CAS-Nummer: 1004192-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR