1-{[3-(Trifluoromethyl)phenoxy]methyl}-1H-pyrazole-3-carboxylic acid
Lieferant: Apollo Scientific
PC410311-1G
APOSPC410311-1GEA
232
EUR
APOSPC410311-1G
1-{[3-(Trifluoromethyl)phenoxy]methyl}-1H-pyrazole-3-carboxylic acid
1-{[3-(Trifluoromethyl)phenoxy]methyl}-1H-pyrazole-3-carboxylic acid
Formel:
C₁₂H₉F₃N₂O₃ Molecular Weight: 286,207 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04967890 CAS-Nummer: 1004193-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...