1-[(3-Chloro-4-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

PC410312-1G
APOSPC410312-1GEA 232 EUR
APOSPC410312-1G
1-[(3-Chloro-4-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
1-[(3-Chloro-4-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
Formel: C₁₁H₈ClFN₂O₃
Molecular Weight: 270,644 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04967901
CAS-Nummer: 1004193-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR