1-[(4-Fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

PC410313-1G
APOSPC410313-1GEA 386 EUR
APOSPC410313-1G
1-[(4-Fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
1-[(4-Fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
Formel: C₁₁H₉FN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04967923
CAS-Nummer: 1004193-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR