1-{[3-(Trifluoromethyl)phenoxy]methyl}-1H-pyrazole-3-carbohydrazide
Lieferant: Apollo Scientific
PC410314-1G
APOSPC410314-1GEA
232
EUR
APOSPC410314-1G
1-{[3-(Trifluoromethyl)phenoxy]methyl}-1H-pyrazole-3-carbohydrazide
1-{[3-(Trifluoromethyl)phenoxy]methyl}-1H-pyrazole-3-carbohydrazide
Formel:
C₁₂H₁₁F₃N₄O₂ Molecular Weight: 300,237 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04967948 CAS-Nummer: 1004193-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...