1-[(3-Chloro-4-fluorophenoxy)methyl]-1H-pyrazole-3-carbohydrazide

Lieferant: Apollo Scientific

PC410315-1G
APOSPC410315-1GEA 232 EUR
APOSPC410315-1G
1-[(3-Chloro-4-fluorophenoxy)methyl]-1H-pyrazole-3-carbohydrazide
1-[(3-Chloro-4-fluorophenoxy)methyl]-1H-pyrazole-3-carbohydrazide
Formel: C₁₁H₁₀ClFN₄O₂
Molecular Weight: 284,674 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04967961
CAS-Nummer: 1004193-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR