1-[(4-Fluorophenoxy)methyl]-1H-pyrazole-3-carbohydrazide
Lieferant: Apollo Scientific
PC410316-1G
APOSPC410316-1GEA
516
EUR
APOSPC410316-1G
1-[(4-Fluorophenoxy)methyl]-1H-pyrazole-3-carbohydrazide
1-[(4-Fluorophenoxy)methyl]-1H-pyrazole-3-carbohydrazide
Formel:
C₁₁H₁₁FN₄O₂ Molecular Weight: 250,229 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04967983 CAS-Nummer: 1004194-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...