4-Chloro-1-[(4-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

PC410317-1G
APOSPC410317-1GEA 232 EUR
APOSPC410317-1G
4-Chloro-1-[(4-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
4-Chloro-1-[(4-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
Formel: C₁₁H₈ClFN₂O₃
Molecular Weight: 270,644 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04968047
CAS-Nummer: 1004194-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR