4-Chloro-1-[3-(trifluoromethyl)benzyl]-1H-pyrazol-3-amine

Lieferant: Apollo Scientific

PC410318-1G
APOSPC410318-1GEA 0 EUR
APOSPC410318-1G
4-Chloro-1-[3-(trifluoromethyl)benzyl]-1H-pyrazol-3-amine
4-Chloro-1-[3-(trifluoromethyl)benzyl]-1H-pyrazol-3-amine
Formel: C₁₁H₉ClF₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02254014
CAS-Nummer: 1004452-00-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR