2-[4-Chloro-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide

Lieferant: Apollo Scientific

PC410319-1G
APOSPC410319-1GEA 516 EUR
APOSPC410319-1G
2-[4-Chloro-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
2-[4-Chloro-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
Formel: C₉H₁₁ClF₂N₄O
Molecular Weight: 264,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04969906
CAS-Nummer: 1004643-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR