1-(Difluoromethyl)-5-methyl-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

PC410323-1G
APOSPC410323-1GEA 237 EUR
APOSPC410323-1G
1-(Difluoromethyl)-5-methyl-1H-pyrazole-3-carboxylic acid
1-(Difluoromethyl)-5-methyl-1H-pyrazole-3-carboxylic acid
Formel: C₆H₆F₂N₂O₂
Molecular Weight: 176.121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04970128
CAS-Nummer: 1004643-64-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR