1-Ethyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
Lieferant: Apollo Scientific
PC410324-1G
APOSPC410324-1GEA
516
EUR
APOSPC410324-1G
1-Ethyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
1-Ethyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
Formel:
C₇H₉F₃N₄O Molecular Weight: 222,168 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970206 CAS-Nummer: 1004643-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...