1-Ethyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

PC410325-1G
APOSPC410325-1GEA 828 EUR
APOSPC410325-1G
1-Ethyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
1-Ethyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
Formel: C₇H₇F₃N₂O₂
Molecular Weight: 208,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04970207
CAS-Nummer: 1004643-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR