2-Hydrazinyl-4-(1-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidine

Lieferant: Apollo Scientific

PC410326-1G
APOSPC410326-1GEA 647 EUR
APOSPC410326-1G
2-Hydrazinyl-4-(1-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidine
2-Hydrazinyl-4-(1-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidine
Formel: C₉H₉F₃N₆
Molecular Weight: 258,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04970346
CAS-Nummer: 1004644-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR