4-(1,3-Dimethyl-1H-pyrazol-4-yl)-2-hydrazinyl-6-(trifluoromethyl)pyrimidine

Lieferant: Apollo Scientific

PC410327-1G
APOSPC410327-1GEA 647 EUR
APOSPC410327-1G
4-(1,3-Dimethyl-1H-pyrazol-4-yl)-2-hydrazinyl-6-(trifluoromethyl)pyrimidine
4-(1,3-Dimethyl-1H-pyrazol-4-yl)-2-hydrazinyl-6-(trifluoromethyl)pyrimidine
Formel: C₁₀H₁₁F₃N₆
Molecular Weight: 272,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04970347
CAS-Nummer: 1004644-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR