4-(1-Ethyl-1H-pyrazol-4-yl)-2-hydrazinyl-6-(trifluoromethyl)pyrimidine
Lieferant: Apollo Scientific
PC410328-1G
APOSPC410328-1GEA
647
EUR
APOSPC410328-1G
4-(1-Ethyl-1H-pyrazol-4-yl)-2-hydrazinyl-6-(trifluoromethyl)pyrimidine
4-(1-Ethyl-1H-pyrazol-4-yl)-2-hydrazinyl-6-(trifluoromethyl)pyrimidine
Formel:
C₁₀H₁₁F₃N₆ Molecular Weight: 272,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970349 CAS-Nummer: 1004644-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...