4-(Difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine
Lieferant: Apollo Scientific
PC410329-1G
APOSPC410329-1GEA
516
EUR
APOSPC410329-1G
4-(Difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine
4-(Difluoromethyl)-6-(1,3-dimethyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine
Formel:
C₁₀H₁₂F₂N₆ Molecular Weight: 254,239 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970384 CAS-Nummer: 1004644-06-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...