4-(Difluoromethyl)-6-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine

Lieferant: Apollo Scientific

PC410330-1G
APOSPC410330-1GEA 516 EUR
APOSPC410330-1G
4-(Difluoromethyl)-6-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine
4-(Difluoromethyl)-6-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-2-hydrazinylpyrimidine
Formel: C₁₁H₁₄F₂N₆
Molecular Weight: 268,266 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04970388
CAS-Nummer: 1004644-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR