2-[5-Cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide

Lieferant: Apollo Scientific

PC410331-1G
APOSPC410331-1GEA 516 EUR
APOSPC410331-1G
2-[5-Cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
2-[5-Cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
Formel: C₉H₁₂F₂N₄O
Molecular Weight: 230.215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04971139
CAS-Nummer: 1004644-16-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR