1-(Difluoromethyl)-5-methyl-1H-pyrazole-3-carbohydrazide
Lieferant: Apollo Scientific
PC410332-1G
APOSPC410332-1GEA
516
EUR
APOSPC410332-1G
1-(Difluoromethyl)-5-methyl-1H-pyrazole-3-carbohydrazide
1-(Difluoromethyl)-5-methyl-1H-pyrazole-3-carbohydrazide
Formel:
C₆H₈F₂N₄O Molecular Weight: 190.151 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04971155 CAS-Nummer: 1004644-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...