2-[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
Lieferant: Apollo Scientific
PC410333-1G
APOSPC410333-1GEA
386
EUR
APOSPC410333-1G
2-[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
2-[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
Formel:
C₇H₈BrF₃N₄O Molecular Weight: 301,064 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419673 CAS-Nummer: 1004644-67-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...