5-(1,3-Dimethyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Lieferant: Apollo Scientific

PC410334-1G
APOSPC410334-1GEA 647 EUR
APOSPC410334-1G
5-(1,3-Dimethyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-2-carboxylic acid
5-(1,3-Dimethyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Formel: C₁₃H₁₄F₃N₅O₂
Molecular Weight: 329.278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03420226
CAS-Nummer: 1005560-38-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR