5-{[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid

Lieferant: Apollo Scientific

PC410335-1G
APOSPC410335-1GEA 190 EUR
APOSPC410335-1G
5-{[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid
5-{[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid
Formel: C₁₁H₈F₄N₂O₃
Molecular Weight: 292,186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419727
CAS-Nummer: 1005565-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR