3-Amino-4-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide

Lieferant: Apollo Scientific

PC410336-1G
APOSPC410336-1GEA 516 EUR
APOSPC410336-1G
3-Amino-4-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide
3-Amino-4-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide
Formel: C₁₅H₁₄F₃N₅OS
Molecular Weight: 369,365 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422770
CAS-Nummer: 1005566-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR