2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Lieferant: Apollo Scientific
PC410337-1G
APOSPC410337-1GEA
386
EUR
APOSPC410337-1G
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel:
C₈H₉F₃N₂O₂ Molecular Weight: 222,164 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419687 CAS-Nummer: 1005566-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...