1,3-Dimethyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
Lieferant: Apollo Scientific
PC410338-1G
APOSPC410338-1GEA
232
EUR
APOSPC410338-1G
1,3-Dimethyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
1,3-Dimethyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
Formel:
C₆H₆F₃N₃O₂ Molecular Weight: 209,126 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970017 CAS-Nummer: 1005576-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...