3-Amino-6-(difluoromethyl)-4-(1-ethyl-5-methyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide

Lieferant: Apollo Scientific

PC410339-1G
APOSPC410339-1GEA 516 EUR
APOSPC410339-1G
3-Amino-6-(difluoromethyl)-4-(1-ethyl-5-methyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
3-Amino-6-(difluoromethyl)-4-(1-ethyl-5-methyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
Formel: C₁₅H₁₅F₂N₅OS
Molecular Weight: 351,374 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422787
CAS-Nummer: 1005582-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR