5-{[4-Chloro-3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid
Lieferant: Apollo Scientific
PC410341-1G
APOSPC410341-1GEA
232
EUR
APOSPC410341-1G
5-{[4-Chloro-3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid
5-{[4-Chloro-3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}furan-2-carboxylic acid
Formel:
C₁₁H₇ClF₄N₂O₃ Molecular Weight: 326,631 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419728 CAS-Nummer: 1005583-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...