2-[4-Chloro-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid
Lieferant: Apollo Scientific
PC410342-1G
APOSPC410342-1GEA
190
EUR
APOSPC410342-1G
2-[4-Chloro-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid
2-[4-Chloro-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid
Formel:
C₈H₉ClF₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419700 CAS-Nummer: 1005584-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...