2-[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Lieferant: Apollo Scientific

PC410343-1G
APOSPC410343-1GEA 516 EUR
APOSPC410343-1G
2-[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
2-[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel: C₈H₈BrF₃N₂O₂
Molecular Weight: 301,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419689
CAS-Nummer: 1005584-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR