2-[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Lieferant: Apollo Scientific

PC410344-1G
APOSPC410344-1GEA 647 EUR
APOSPC410344-1G
2-[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]propanoic acid
2-[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel: C₈H₈F₄N₂O₂
Molecular Weight: 240,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419715
CAS-Nummer: 1005586-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR